Assay Details
Gene Symbol: 2810402E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006567.1 | - | 1 | 347 | 96 | |
AK012965.1 | - | 1 | 293 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ubiquitin specific peptidase 54 | Usp54 |
4930429G18Rik, AI115571, C030002J06Rik |
Anti-sense | Chr. 14 - 20548912 - 20643129 on NCBI Build GRCm38 | |
Downstream | myozenin 1 | Myoz1 |
2310001N11Rik, AV090278, FATZ, Myoz |
7.6kb | Anti-sense | Chr. 14 - 20649102 - 20656540 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
