Assay Details
Gene Symbol: 1700064F23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK006880.1 | - | 1 - 2 | 216 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tyrosine aminotransferase | Tat | -7.1kb | Sense | Chr. 8 - 109990436 - 109999804 on NCBI Build GRCm38 | |
Downstream | carbohydrate (chondroitin 6/keratan) sulfotransferase 4 | Chst4 |
GST-3, Gn6st-2, HEC-GlcNAc6ST |
21.4kb | Anti-sense | Chr. 8 - 110029075 - 110039375 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
