Assay Details
Gene Symbol: 2310001K24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_028122.1 | 1 | 349 | 78 | ||
GenBank mRNA | AK009071.1 | - | 1 | 352 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | fat storage-inducing transmembrane protein 2 | Fitm2 |
AI414861, D930001I22Rik, Fit2 |
Anti-sense | Chr. 2 - 163468703 - 163472629 on NCBI Build GRCm38 | |
Downstream | R3H domain containing-like | R3hdml | OTTMUSG00000001070 | 18.2kb | Sense | Chr. 2 - 163491214 - 163504364 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
