Assay Details
Gene Symbol: A930011B18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK020844.1 | - | 1 | 643 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin-dependent kinase 1 | Cdk1 |
Cdc2, Cdc2a, p34 |
-86.1kb | Sense | Chr. 10 - 69336635 - 69352912 on NCBI Build GRCm38 |
Downstream |
ankyrin 3, epithelial |
Ank3 |
2900054D09Rik, AI314020, Ank-3, AnkG, Ankyrin-3, Ankyrin-G |
93.8kb | Anti-sense | Chr. 10 - 69533580 - 70027436 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
