Assay Details
Gene Symbol: Rapgef6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021094.1 | - | 1 | 650 | 85 | |
AK038235.1 | - | 1 | 85 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | folliculin interacting protein 1 | Fnip1 |
A730024A03Rik, AI838773, AW557298 |
-78.5kb | Sense | Chr. 11 - 54438146 - 54518241 on NCBI Build GRCm38 |
Downstream | CDC42 small effector 2 | Cdc42se2 |
2810404F18Rik, AA408783, AA536669, SPEC2 |
119.9kb | Anti-sense | Chr. 11 - 54717415 - 54787703 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
