Assay Details
Gene Symbol: D530033B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK021305.1 | - | 1 | 148 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cyclin E1 | Ccne1 |
AW538188, CycE1 |
-75.1kb | Sense | Chr. 7 - 38097984 - 38107490 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1600014C10 gene | 1600014C10Rik | AI428873 | 439.0 | Anti-sense | Chr. 7 - 38183217 - 38197568 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
