Assay Details
Gene Symbol: E230016M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040278.1 | 1 - 2 | 341 | 91 | ||
GenBank mRNA | AK029473.1 | - | 1 - 2 | 342 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
guanine nucleotide binding protein (G protein), gamma 12 |
Gng12 |
2010305F15Rik, AA536815, AI115529, AI314170, AI842738 |
-15.2kb | Sense | Chr. 6 - 66896397 - 67021361 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA A430010J10 gene | A430010J10Rik | 94.3kb | Sense | Chr. 6 - 67157801 - 67169010 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
