Assay Details
Gene Symbol: E230016M11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040278.1 | 5 - 6 | 821 | 96 | ||
GenBank mRNA | AK029473.1 | - | 5 - 6 | 822 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | growth arrest and DNA-damage-inducible 45 alpha | Gadd45a |
AA545191, Ddit1, GADD45 |
-39.8kb | Anti-sense | Chr. 6 - 67035096 - 67037407 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA A430010J10 gene | A430010J10Rik | 79.4kb | Sense | Chr. 6 - 67157801 - 67169010 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
