Assay Details
Gene Symbol: 4930417O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK030803.1 | - | 102 | 92 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA E130112N10 gene | E130112N10Rik |
5830448L21Rik, 9530059J11Rik |
-25.5kb | Sense | Chr. 6 - 125231276 - 125239934 on NCBI Build GRCm38 |
Downstream |
tubulin, alpha 3A |
Tuba3a |
AI528779, M[a]3, Tuba3 |
11.7kb | Anti-sense | Chr. 6 - 125278274 - 125286042 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
