Assay Details
Gene Symbol: A230001M10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040391.1 | 2 - 3 | 640 | 91 | ||
GenBank mRNA | AK038444.1 | - | 2 - 3 | 524 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
vang-like 1 (van gogh, Drosophila) |
Vangl1 |
Kitenin, Lpp2, Stb2, Stbm |
-63.1kb | Anti-sense | Chr. 3 - 102155899 - 102205011 on NCBI Build GRCm38 |
Downstream | nerve growth factor | Ngf | Ngfb | 190.6kb | Sense | Chr. 3 - 102469919 - 102521013 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2374381 | BB169751.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2374381 | AK038444.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 611604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 917089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 451377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 354286 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 669428 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 322660 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 182547 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 99636 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 115412 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 11824 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 630928 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 399360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300784 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 724530 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 748355 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467119 | BC119059.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467119 | BC125619.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 467119 | NM_146702.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289724 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 243016 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 951169 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 408326 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 79130 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 578330 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229027 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 513346 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300365 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215041 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 871956 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 606181 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 218334 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 966750 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227997 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 880700 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 208356 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 532387 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712360 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22661 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831539 | XM_001479157.1 | refseq_mrna_mouse_XM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 526526 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 618421 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 829914 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 195908 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 811248 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852097 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 106196 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 490316 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 90053 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 122225 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 310148 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 380735 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 111717 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 456359 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 575894 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 663154 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 892028 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525718 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 309141 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 9324 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 183524 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 807089 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 696503 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 474569 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23240 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 36885 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 118144 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 76397 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 101658 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 99769 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 55406 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16487 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119377 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 157978 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 74190 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P132269 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2374381 | BB169751.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2374381 | AK038444.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P132269 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011061 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011227 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05010949 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011159 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05010983 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05010969 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00713555 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011223 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011183 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011079 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011285 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011027 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00713657 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P05011295 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101226 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05011079 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05011227 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P132269 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05011159 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P05011285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380018 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02970633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02982882 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380904 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380576 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02795032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380167 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02672683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02722667 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380981 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380012 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380155 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380943 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380967 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380930 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380064 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380950 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02795054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380127 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02414122 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03172595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380061 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380161 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380032 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02372566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02446683 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02294412 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02446693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380053 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02410144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380946 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02309026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380029 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380893 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380892 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380275 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380937 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380976 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380093 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380268 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380273 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03113069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02420168 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380086 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02385960 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380240 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380082 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380948 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02372568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380163 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02762999 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380908 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380291 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380285 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380003 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380239 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380898 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02376480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380080 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380038 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02393355 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380236 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03180569 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380980 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380087 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02414124 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02294413 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02499179 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380935 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02384900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02414126 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02294414 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380100 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380257 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380060 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03154772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02446696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02499183 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02385962 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02795031 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02689109 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02343430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02376479 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02722668 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380069 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380880 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380022 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380040 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02795042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02483825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380079 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380049 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02746453 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380900 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380945 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02502512 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380912 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380006 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02309039 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380266 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380084 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380136 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380952 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03067544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380055 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380910 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380883 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02384899 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380261 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380065 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02309030 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03113072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02755337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02376477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380037 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03169720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02746455 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03064339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02672684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380903 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02691283 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380971 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380913 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02343431 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380573 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380095 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03180570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380940 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380072 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380096 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02795051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380158 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380260 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03064337 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03019466 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02687501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380047 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380142 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02945902 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02502511 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03008591 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380977 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380957 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380278 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380256 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380005 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380141 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22380014 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1457570_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1445715_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1457570_at | - | - |
Mouse Genome 430 2.0 Array | 1457570_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445715_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1445715_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300008254.mm.1 | TC0300001028.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300008254.mm.1 | NR_040391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008254.mm.1 | NR_040391.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008256.mm.1 | TC0300001028.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300008256.mm.1 | NR_040391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008256.mm.1 | NR_040391.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008260.mm.1 | TC0300001028.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300008260.mm.1 | NR_040391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008260.mm.1 | NR_040391.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008259.mm.1 | TC0300001028.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300008259.mm.1 | NR_040391.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300008259.mm.1 | NR_040391.1 | Refseq |
Mouse Expression Set 430 | 1445715_at | - | - |
Mouse Expression Set 430 | 1445715_PM_at | - | - |
Mouse Expression Set 430 | 1457570_at | - | - |
Mouse Expression Set 430 | 1457570_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445715_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1445715_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457570_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1457570_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 548298 | - | - |
