Assay Details
Gene Symbol: Mgea5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK040197.1 | - | 54 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | nucleoplasmin 3 | Npm3 | Nub1 | -63.0 | Anti-sense | Chr. 19 - 45747734 - 45749563 on NCBI Build GRCm38 |
Downstream | Kv channel-interacting protein 2 | Kcnip2 | KChIP2 | 40.0kb | Anti-sense | Chr. 19 - 45792346 - 45816063 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
