Assay Details
Gene Symbol: A730080H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK047900.1 | - | 1 | 1050 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BU513811.1 | - | 160 | Chr. - - [-] on Build | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 352 pseudogene | Gm12633 | OTTMUSG00000007790 | -850.2kb | Sense | Chr. 4 - 90359625 - 90361314 on NCBI Build GRCm38 |
Downstream |
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 2 (Hu antigen B) |
Elavl2 |
Hub, mel-N1 |
37.0kb | Sense | Chr. 4 - 91250766 - 91400064 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
