Assay Details
Gene Symbol: 4930470O06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK015539.1 | - | 3 - 4 | 440 | 163 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | cDNA sequence BC030500 | BC030500 | -915.1kb | Sense | Chr. 8 - 58911598 - 58914298 on NCBI Build GRCm38 | |
Downstream | aminoadipate aminotransferase | Aadat |
AI875679, Aadt, KATII, Kat2, mKat-2 |
661.1kb | Sense | Chr. 8 - 60505870 - 60545677 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
