Assay Details
Gene Symbol: 4933423L19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK016876.1 | - | 1 | 1343 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4930402F11 gene | 4930402F11Rik | -449.2kb | Sense | Chr. 7 - 69496094 - 69499689 on NCBI Build GRCm38 | |
Downstream |
predicted gene, 29683 |
Gm29683 | 317.3kb | Anti-sense | Chr. 7 - 70267693 - 70339083 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 516532 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208178 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208150 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208227 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06496212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208191 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208166 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06187369 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208200 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06206656 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208175 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208107 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06237600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208193 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208112 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06267690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208222 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208229 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208180 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208195 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208212 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208230 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208220 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208206 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208115 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208153 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208121 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208151 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208168 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208130 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208234 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208218 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208126 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208164 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208108 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06206655 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208162 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208177 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208123 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208205 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208154 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06187367 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208232 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208233 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208223 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P05997538 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208216 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208131 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208224 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208118 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208184 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208165 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208213 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06069600 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06187361 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208190 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208176 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208132 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208163 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208125 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208221 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208198 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208127 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208156 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208203 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208196 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208192 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208120 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P25208183 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1454459_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1433061_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1433061_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1454459_at | - | - |
Mouse Genome 430 2.0 Array | 1433061_at | - | - |
Mouse Genome 430 2.0 Array | 1454459_PM_at | - | - |
Mouse Expression Set 430 | 1454459_at | - | - |
Mouse Expression Set 430 | 1454459_PM_at | - | - |
Mouse Expression Set 430 | 1433061_PM_at | - | - |
Mouse Expression Set 430 | 1433061_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433061_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454459_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1433061_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1454459_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 101170441 | - | - |
