Assay Details
Gene Symbol: 4930502C15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK076703.1 | - | 305 | 71 | ||
AK076827.1 | - | 656 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
C-type lectin domain family 2, member L |
Clec2l |
AI414054, AW122724, Gm1064 |
-60.7kb | Sense | Chr. 6 - 38663069 - 38680865 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
