Assay Details
Gene Symbol: Gm20556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040347.1 | 1 - 2 | 279 | 101 | ||
GenBank mRNA | AK079813.1 | - | 1 - 2 | 280 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | proline rich 5 (renal) | Prr5 |
AU043908, Arhgap8, C030017C09Rik, C78947, Protor-1, Protor1 |
-12.3kb | Anti-sense | Chr. 15 - 84663171 - 84706409 on NCBI Build GRCm38 |
Downstream | PHD finger protein 21B | Phf21b | A730032D07Rik | 65.1kb | Sense | Chr. 15 - 84785376 - 84856132 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
