Assay Details
Gene Symbol: Rdx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK085332.1 | - | 1 | 1052 | 83 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | glyceraldehyde-3-phosphate dehydrogenase pseudogene | Gm6981 | EG629557 | -23.1kb | Anti-sense | Chr. 9 - 52002061 - 52048705 on NCBI Build GRCm38 |
Downstream | zinc finger CCCH type containing 12C | Zc3h12c |
A230108E06, C230027N18Rik, mKIAA1726 |
36.4kb | Anti-sense | Chr. 9 - 52109583 - 52169154 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
