Assay Details
Gene Symbol: Gm15133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040749.1 | 1 - 2 | 76 | 103 | ||
GenBank mRNA | AK085986.1 | - | 1 - 2 | 77 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | tripartite motif-containing 5 | Trim5 |
EG667823, Gm8833 |
-26.6kb | Anti-sense | Chr. 7 - 104263386 - 104288421 on NCBI Build GRCm38 |
Downstream | tripartite motif-containing 34B | Trim34b |
Gm15134, OTTMUSG00000019188, Trim34-2 |
12.1kb | Sense | Chr. 7 - 104329471 - 104336617 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
