Assay Details
Gene Symbol: D030024E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040350.1 | 1 - 2 | 71 | 90 | ||
GenBank mRNA | AK132958.1 | - | 1 - 2 | 73 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 29904 |
Gm29904 | -441.4kb | Anti-sense | Chr. 15 - 61288441 - 61331370 on NCBI Build GRCm38 | |
Downstream | myelocytomatosis oncogene | Myc |
AU016757, Myc2, Niard, Nird, bHLHe39 |
210.9kb | Anti-sense | Chr. 15 - 61985341 - 61990361 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
