Assay Details
Gene Symbol: Gm5833 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040304.1 | 2 - 3 | 218 | 96 | ||
GenBank mRNA | AK154631.1 | - | 2 - 3 | 220 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NIMA (never in mitosis gene a)-related expressed kinase 7 | Nek7 |
2810460C19Rik, AU020186 |
-37.0kb | Sense | Chr. 1 - 138484106 - 138619779 on NCBI Build GRCm38 |
Downstream | LIM homeobox protein 9 | Lhx9 |
3110009O07Rik, BB104635, LH2B |
147.3kb | Sense | Chr. 1 - 138825186 - 138848649 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

This assay targets the same location as RTA transcript cluster(s) TC1300001516.rn.1
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 972783 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 18695 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 189467 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 621587 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291634 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579054 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 101409 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 550209 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 457116 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 561534 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 686519 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 102929 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 98155 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 284674 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 65554 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 129655 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 108476 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P123655 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P1977895 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04282485 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04282464 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P123655 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P123655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658609 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658656 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00806209 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658737 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658649 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658544 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658551 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658597 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658557 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658505 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658669 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658570 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658695 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658575 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658594 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00505501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00639820 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658664 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658574 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658676 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658539 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658543 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658617 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658638 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00652965 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658481 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658684 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658495 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658710 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658604 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658722 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658693 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00672886 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00806216 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658674 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658715 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658507 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658586 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658729 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00656517 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658606 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658632 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658566 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658615 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01037645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658595 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01008353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658720 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00360612 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658550 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658554 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01008354 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01068209 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00656521 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658496 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658477 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658647 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658562 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658689 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658712 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658560 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658486 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658603 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658475 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658494 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658498 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00305526 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658608 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658750 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658564 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658640 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658636 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00269884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658497 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00339228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658545 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658745 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658568 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658587 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658744 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658652 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658713 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00166186 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658559 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658634 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658728 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658651 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00553696 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00672884 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658473 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658686 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658583 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658633 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00142798 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658699 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00656536 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658654 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658653 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658488 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658682 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00467538 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658490 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658706 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658730 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658561 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658580 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658677 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658571 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00045365 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658727 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658484 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658607 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658631 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658691 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01130480 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658666 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658726 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658500 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658655 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P20658756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P00656524 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0100025737.mm.1 | TC0100003173.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100025737.mm.1 | NR_040304.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100025737.mm.1 | NR_040304.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100025736.mm.1 | TC0100003173.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100025736.mm.1 | NR_040304.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100025736.mm.1 | NR_040304.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100025738.mm.1 | TC0100003173.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0100025738.mm.1 | NR_040304.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0100025738.mm.1 | NR_040304.1 | Refseq |
