Assay Details
Gene Symbol: Cdc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK012511.1 | - | 1 | 1046 | 70 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 4932443I19 gene | 4932443I19Rik |
A230061C02Rik, Gm1106 |
-36.3kb | Sense | Chr. 8 - 13705449 - 13743531 on NCBI Build GRCm38 |
Downstream | UPF3 regulator of nonsense transcripts homolog A (yeast) | Upf3a |
2600001C03Rik, 4930546M19Rik, RENT3A, UPF3 |
4.0kb | Sense | Chr. 8 - 13785215 - 13798538 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
