Assay Details
Gene Symbol: 2900040J22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013628.1 | - | 1 | 942 | 100 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | CBP80/20-dependent translation initiation factor | Ctif | Gm672 | Sense | Chr. 18 - 75431205 - 75697696 on NCBI Build GRCm38 | |
Downstream | predicted gene 10532 | Gm10532 | ENSMUSG00000073534 | 43.9kb | Anti-sense | Chr. 18 - 75514645 - 75522806 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
