Assay Details
Gene Symbol: 2900092N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK013842.1 | - | 1 | 146 | 116 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
glutamate receptor, ionotropic, AMPA2 (alpha 2) |
Gria2 |
GluA2, GluR-B, Glur-2, Glur2 |
-40.0kb | Sense | Chr. 3 - 80684935 - 80803204 on NCBI Build GRCm38 |
Downstream |
glycine receptor, beta subunit |
Glrb |
AI853901, Glyrb, spa, spastic |
51.0 | Sense | Chr. 3 - 80843598 - 80913660 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
