Assay Details
Gene Symbol: 0910001E24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK003069.1 | - | 1 | 145 | 111 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 12514 | Gm12514 | OTTMUSG00000007237 | -152.4kb | Anti-sense | Chr. 4 - 55129714 - 55139159 on NCBI Build GRCm38 |
Downstream | RAD23b homolog (S. cerevisiae) | Rad23b |
0610007D13Rik, AV001138, HR23B, mHR23B, p58 |
58.2kb | Sense | Chr. 4 - 55350042 - 55392237 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
