Assay Details
Gene Symbol: Klhdc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004455.1 | - | 1 | 650 | 93 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | prefoldin 2 | Pfdn2 |
ESTM27, W48336 |
-235.0 | Sense | Chr. 1 - 171345699 - 171358170 on NCBI Build GRCm38 |
Downstream | poliovirus receptor-related 4 | Pvrl4 |
1200017F15Rik, Prr4 |
10.9kb | Sense | Chr. 1 - 171370173 - 171388287 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
