Assay Details
Gene Symbol: 4930588D02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK019831.1 | - | 1 | 247 | 94 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mannoside acetylglucosaminyltransferase 3 | Mgat3 |
1110038J12Rik, AI848272, GnT-III |
-12.4kb | Anti-sense | Chr. 15 - 80173650 - 80215519 on NCBI Build GRCm38 |
Downstream | mitochondrial elongation factor 1 | Mief1 |
A230016E22, AI452372, Smcr7l |
5.5kb | Anti-sense | Chr. 15 - 80233940 - 80253373 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
