Assay Details
Gene Symbol: Gm33677 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK132971.1 | - | 3 - 4 | 459 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
predicted gene, 38403 |
Gm38403 | C230029M16 | -101.1kb | Sense | Chr. 10 - 118920381 - 118927143 on NCBI Build GRCm38 |
Downstream | RIKEN cDNA 1700064J06 gene | 1700064J06Rik | 48.5kb | Anti-sense | Chr. 10 - 119092905 - 119103107 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
