Assay Details
Gene Symbol: LOC105244170 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK138581.1 | - | 6 - 7 | 1174 | 102 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | neuromedin U receptor 2 | Nmur2 | Nmu2r | -32.9kb | Anti-sense | Chr. 11 - 56024990 - 56040987 on NCBI Build GRCm38 |
Downstream |
glutamate receptor, ionotropic, AMPA1 (alpha 1) |
Gria1 |
2900051M01Rik, Glr-1, Glr1, GluA1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, gluR-K1 |
936.0kb | Sense | Chr. 11 - 57011571 - 57330244 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
