Assay Details
Gene Symbol: Gm40761 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK142676.1 | - | 1 - 2 | 236 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | Bardet-Biedl syndrome 10 (human) | Bbs10 |
1300007O09Rik, AI452285 |
-35.4kb | Sense | Chr. 10 - 111298679 - 111301736 on NCBI Build GRCm38 |
Downstream | nucleosome assembly protein 1-like 1 | Nap1l1 |
AA407126, AI256722, D10Ertd68e, NAP-1 |
129.2kb | Sense | Chr. 10 - 111473192 - 111498150 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
