Assay Details
Gene Symbol: Gm12100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK049252.1 | - | 344 | 64 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
acylphosphatase 2, muscle type |
Acyp2 | 2310004B09Rik | -101.4kb | Anti-sense | Chr. 11 - 30505992 - 30649396 on NCBI Build GRCm38 |
Downstream |
proteasome (prosome, macropain) activator subunit 4 |
Psme4 |
AA409398, AU041366, TEMO, mKIAA0077 |
16.5kb | Sense | Chr. 11 - 30771328 - 30881150 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
