Assay Details
Gene Symbol: Gm15558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134234.1 | - | 4 - 5 | 1131 | 78 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | dexamethasone-induced transcript | Dexi |
1810029J14Rik, AI836170, AW413143, D16Bwg0586e, Myle |
-126.1kb | Sense | Chr. 16 - 10530207 - 10543122 on NCBI Build GRCm38 |
Downstream | suppressor of cytokine signaling 1 | Socs1 |
Cish1, Cish7, JAB, SOCS-1, SSI-1 |
111.9kb | Sense | Chr. 16 - 10783808 - 10785536 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
