Assay Details
Gene Symbol: Gm15326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK134426.1 | - | 1 - 2 | 146 | 61 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | mitogen-activated protein kinase kinase kinase 1 | Map3k1 |
MAPKKK1, MEKK1, Mekk |
-49.7kb | Anti-sense | Chr. 13 - 111746433 - 111818269 on NCBI Build GRCm38 |
Downstream | ankyrin repeat domain 55 | Ankrd55 | C030011J08Rik | 418.3kb | Sense | Chr. 13 - 112288451 - 112384002 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
