Assay Details
Gene Symbol: Gm31121 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK139864.1 | - | 3 - 4 | 367 | 65 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | adhesion G protein-coupled receptor L2 | Adgrl2 |
AI450192, Gm619, Lec1, Lphh1, Lphn2, mKIAA0786 |
-126.3kb | Sense | Chr. 3 - 148815586 - 149445423 on NCBI Build GRCm38 |
Downstream | microRNA 3963 | Mir3963 |
mmu-mir-3963, mu-mir-3963 |
1.4mb | Sense | Chr. 3 - 151023792 - 151023862 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
