Assay Details
Gene Symbol: Gm19710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045749.1 | 2 - 3 | 319 | 101 | ||
GenBank mRNA | AK140585.1 | - | 2 - 3 | 321 | 101 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | HCLS1 associated X-1 | Hax1 |
HAX-1, HSP1BP-1, Hs1bp1, SIG-111, Silg111, mHAX-1s |
-429.0 | Anti-sense | Chr. 3 - 89995446 - 89998716 on NCBI Build GRCm38 |
Downstream | microRNA 7669 | Mir7669 |
Gm27996, mmu-mir-7669 |
10.9kb | Anti-sense | Chr. 3 - 90011153 - 90011210 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
