Assay Details
Gene Symbol: Gm31774 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143772.1 | - | 1 - 2 | 217 | 81 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | hydroxysteroid (17-beta) dehydrogenase 2 | Hsd17b2 |
AI194836, AI194967, AI255511 |
Anti-sense | Chr. 8 - 117700982 - 117759030 on NCBI Build GRCm38 | |
Downstream | M phase phosphoprotein 6 | Mphosph6 |
1110001M01Rik, AA536809, C86426 |
43.4kb | Sense | Chr. 8 - 117791642 - 117801933 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
