Assay Details
Gene Symbol: Gm30025 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK147035.1 | - | 1 | 651 | 80 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | small Cajal body-specific RNA 3B | Scarna3b | MBI-100 | -43.5kb | Anti-sense | Chr. 12 - 80391614 - 80391737 on NCBI Build GRCm38 |
Downstream | exonuclease 3'-5' domain containing 2 | Exd2 |
4930539P14Rik, C85658, Exdl2 |
27.2kb | Sense | Chr. 12 - 80463095 - 80498135 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
