Assay Details
Gene Symbol: Gm35156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK156776.1 | - | 2 - 3 | 91 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | EF-hand domain (C-terminal) containing 2 | Efhc2 |
4933407D04Rik, mRib72-2 |
-221.4kb | Anti-sense | Chr. X - 17132049 - 17320461 on NCBI Build GRCm38 |
Downstream | dual specificity phosphatase 21 | Dusp21 |
1700094E07Rik, DSP21 |
591.0kb | Sense | Chr. X - 18145870 - 18146694 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
