Assay Details
Gene Symbol: Gm29461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC072606.1 | - | 2 - 3 | 181 | 77 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 1810006J02 gene | 1810006J02Rik | -30.4kb | Sense | Chr. 1 - 98131467 - 98144655 on NCBI Build GRCm38 | |
Downstream |
solute carrier organic anion transporter family, member 6d1 |
Slco6d1 | 4921511I05Rik | 235.5kb | Sense | Chr. 1 - 98421085 - 98516985 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
