Assay Details
Gene Symbol: Gm35915 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK078274.1 | - | 1 | 751 | 89 | |
AK148359.1 | - | 1 | 770 | 89 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | carbohydrate sulfotransferase 2 | Chst2 |
AI428561, AW121776, C130041E03Rik, Chts2, GST-2, GlcNAc6ST, Gn6st |
-209.0 | Anti-sense | Chr. 9 - 95400926 - 95407270 on NCBI Build GRCm38 |
Downstream | U2 snRNP-associated SURP domain containing | U2surp |
2610101N10Rik, AU023006, Sr140 |
48.3kb | Anti-sense | Chr. 9 - 95456894 - 95513208 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
