Assay Details
Gene Symbol: 5830416I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045384.1 | 3 | 2353 | 86 | ||
GenBank mRNA | AK080054.1 | - | 1 | 744 | 86 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RELT-like 1 | Rell1 | AA536743 | -80.3kb | Anti-sense | Chr. 5 - 63888667 - 63968900 on NCBI Build GRCm38 |
Downstream | phosphoglucomutase 1 | Pgm1 |
3230402E02Rik, Pgm-1, Pgm2 |
42.6kb | Sense | Chr. 5 - 64092831 - 64128355 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
