Assay Details
Gene Symbol: Gm30127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131720.1 | - | 3 - 4 | 583 | 90 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | ribonuclease P 40 subunit | Rpp40 |
D8Bwg1265e, Rnasep1 |
-48.4kb | Sense | Chr. 13 - 35895104 - 35911565 on NCBI Build GRCm38 |
Downstream | LYR motif containing 4 | Lyrm4 |
BC034664, Gm903 |
17.6kb | Sense | Chr. 13 - 35978797 - 36117357 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
