Assay Details
Gene Symbol: Gm12348 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158747.1 | - | 2 - 3 | 195 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | junction plakoglobin | Jup |
Ctnng, D930025P04Rik, PG |
-3.7kb | Anti-sense | Chr. 11 - 100368856 - 100397790 on NCBI Build GRCm38 |
Downstream | prolyl 3-hydroxylase family member 4 (non-enzymatic) | P3h4 |
1110036O03Rik, AI413214, Leprel4, Sc65 |
6.5kb | Anti-sense | Chr. 11 - 100408748 - 100414819 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
