Assay Details
Gene Symbol: Gm26947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158810.1 | - | 4 - 5 | 1211 | 96 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | microRNA 6408 | Mir6408 |
Gm27451, mmu-mir-6408 |
-14.7kb | Sense | Chr. 10 - 60923023 - 60923113 on NCBI Build GRCm38 |
Downstream | predicted gene 20611 | Gm20611 | 4930565A17 | 90.4kb | Sense | Chr. 10 - 61031303 - 61038080 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
