Assay Details
Gene Symbol: Gm16582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158969.1 | - | 1 - 2 | 67 | 71 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
wingless-type MMTV integration site family, member 10A |
Wnt10a | -46.7kb | Sense | Chr. 1 - 74792019 - 74804176 on NCBI Build GRCm38 | |
Downstream |
cyclin-dependent kinase 5, regulatory subunit 2 (p39) |
Cdk5r2 |
B230310J22Rik, p39 |
514.0 | Sense | Chr. 1 - 74855029 - 74857732 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
