Assay Details
Gene Symbol: Gm16582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158969.1 | - | 2 - 3 | 245 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
wingless-type MMTV integration site family, member 10A |
Wnt10a | -50.2kb | Sense | Chr. 1 - 74792019 - 74804176 on NCBI Build GRCm38 | |
Downstream | FEV (ETS oncogene family) | Fev |
Pet-1, Pet1, mPet-1 |
12.7kb | Anti-sense | Chr. 1 - 74881509 - 74885408 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() |
