Assay Details
Gene Symbol: Gm16582 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK158969.1 | - | 3 - 4 | 346 | 73 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cyclin-dependent kinase 5, regulatory subunit 2 (p39) |
Cdk5r2 |
B230310J22Rik, p39 |
-11.0kb | Sense | Chr. 1 - 74855029 - 74857732 on NCBI Build GRCm38 |
Downstream | FEV (ETS oncogene family) | Fev |
Pet-1, Pet1, mPet-1 |
1.6kb | Anti-sense | Chr. 1 - 74881509 - 74885408 on NCBI Build GRCm38 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
