Assay Details
Gene Symbol: Gm9857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK005845.1 | - | 1 | 452 | 91 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | PRP38 pre-mRNA processing factor 38 (yeast) domain containing B | Prpf38b |
1110021E09Rik, AU018955 |
-28.1kb | Sense | Chr. 3 - 108902807 - 108911752 on NCBI Build GRCm38 |
Downstream |
family with sequence similarity 102, member B |
Fam102b |
1600010D10Rik, B430201A12Rik |
30.5kb | Sense | Chr. 3 - 108970997 - 109027607 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
