Assay Details
Gene Symbol: Gm32441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK131795.1 | - | 1 - 2 | 38 | 66 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | NHL repeat containing 2 | Nhlrc2 |
1200003G01Rik, AI835049, AV002846, AW496455 |
-60.1kb | Anti-sense | Chr. 19 - 56548261 - 56603503 on NCBI Build GRCm38 |
Downstream |
adrenergic receptor, beta 1 |
Adrb1 |
Adrb-1, beta-AR |
57.8kb | Anti-sense | Chr. 19 - 56722372 - 56724862 on NCBI Build GRCm38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
