Assay Details
Gene Symbol: Gm13003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040443.1 | 1 - 2 | 1897 | 99 | ||
GenBank mRNA | AK132559.1 | - | 1 - 2 | 1898 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 13001 | Gm13001 | OTTMUSG00000009820 | -43.6kb | Sense | Chr. 4 - 137116705 - 137118255 on NCBI Build GRCm38 |
Downstream |
wingless-type MMTV integration site family, member 4 |
Wnt4 | Wnt-4 | 112.6kb | Anti-sense | Chr. 4 - 137277635 - 137299501 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
