Assay Details
Gene Symbol: Gm13003 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040443.1 | 3 - 4 | 2271 | 109 | ||
GenBank mRNA | AK132559.1 | - | 3 - 4 | 2272 | 109 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | predicted gene 13001 | Gm13001 | OTTMUSG00000009820 | -40.8kb | Sense | Chr. 4 - 137116705 - 137118255 on NCBI Build GRCm38 |
Downstream |
wingless-type MMTV integration site family, member 4 |
Wnt4 | Wnt-4 | 117.5kb | Anti-sense | Chr. 4 - 137277635 - 137299501 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
